Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007789.3 | NP_031815.2 | 15 | 3873 | ||
GenBank mRNA | AK082298.1 | - | 5 | 598 | ||
BC065118.1 | - | 15 | 3839 | 6853253 | ||
X84727.1 | - | 15 | 3835 |