Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dapk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190473.1 | NP_001177402.1 | 3 | 408 | ||
NM_001190474.1 | NP_001177403.2 | 3 | 460 | |||
NM_007828.2 | NP_031854.1 | 2 | 428 | |||
GenBank mRNA | AB007143.1 | - | 2 | 341 | ||
AK035171.1 | - | 1 | 31 | |||
AK158299.1 | - | 3 | 451 | |||
BC012210.1 | - | 3 | 426 | |||
BC023238.1 | - | 3 | 398 | |||
BC125443.1 | - | 2 | 428 | 40129958 | ||
BC137680.1 | - | 2 | 428 | 8860699 |