Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Defa-rs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007845.3 | NP_031871.1 | 148 | |||
GenBank mRNA | BC119615.1 | - | 148 | 40061719 | ||
BC119616.1 | - | 148 | 40061721 |