Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007879.1 | NP_031905.1 | 2 | 180 | ||
GenBank mRNA | AK044194.1 | - | 2 | 189 | ||
AK080814.1 | - | 2 | 178 | |||
AK090156.1 | - | 2 | 210 | |||
AK144438.1 | - | 2 | 182 | |||
AK155061.1 | - | 2 | 195 | |||
AK208688.1 | - | 1 | 56 | |||
BC039649.1 | - | 2 | 174 | 5351885 | ||
D10715.1 | - | 2 | 180 |