Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMTF1 SIRNA Assays ›
Gene Symbol:
DMTF1
Gene Name:
cyclin D binding myb like transcription factor 1
Gene Aliases:
DMP1, DMTF, MRUL, hDMP1
Chromosome Location:
Chr.7: 87152361 - 87196332 on Build GRCh38
UniGene ID:
Hs.196129
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6109
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DMTF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142326.1
NP_001135798.1
10
1240
NM_001142327.1
NP_001135799.1
11
1357
NM_021145.3
NP_066968.3
13
1594
NR_024549.1
11
1568
NR_024550.1
11
1529
XM_011516735.2
XP_011515037.1
11
2279
XM_011516737.1
XP_011515039.1
12
1404
XM_011516739.2
XP_011515041.1
10
1169
XM_017012865.1
XP_016868354.1
12
2373
XM_017012866.1
XP_016868355.1
10
2156
XM_017012867.1
XP_016868356.1
9
2824
XM_017012868.1
XP_016868357.1
12
1404
XM_017012869.1
XP_016868358.1
13
1433
XM_017012870.1
XP_016868359.1
10
1172
XM_017012871.1
XP_016868360.1
12
2545
XM_017012872.1
XP_016868361.1
11
2451
XM_017012873.1
XP_016868362.1
12
2340
XM_017012874.1
XP_016868363.1
12
1576
XM_017012875.1
XP_016868364.1
12
2379
XM_017012876.1
XP_016868365.1
10
1319
XM_017012877.1
XP_016868366.1
11
1438
XM_017012878.1
XP_016868367.1
13
2473
GenBank mRNA
AB209230.1
-
10
1363
AF084530.1
-
11
1306
AF202144.1
-
11
1478
AF202145.1
-
11
1517
AK314622.1
-
11
1293
BC007418.2
-
2
306
BC007447.2
-
2
306
BC070064.1
-
13
1540
4373991
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053693, XM_001060830, XM_006235993, BC081880.This siRNA also targets the following transcripts in Mouse: NM_001110327, NM_011806, AK078015, BC045141, AK046201, and others.
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor recruiting
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0