Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dach1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038610.2 | NP_001033699.1 | 2 | 1306 | ||
NM_007826.3 | NP_031852.1 | 2 | 1306 | |||
GenBank mRNA | AF090436.1 | - | 2 | 850 | ||
AF090437.1 | - | 2 | 850 | |||
AF102547.1 | - | 2 | 850 | |||
AF129510.1 | - | 2 | 1068 | |||
AJ005669.1 | - | 2 | 368 | |||
AK047409.1 | - | 2 | 172 | |||
AK053594.1 | - | 2 | 515 | |||
AK139060.1 | - | 2 | 1307 | |||
BC059934.1 | - | 2 | 1084 | |||
BC078644.1 | - | 2 | 840 | |||
BC141130.1 | - | 2 | 912 | 9055742 | ||
BC145440.1 | - | 2 | 912 |