Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Drg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007879.1 | NP_031905.1 | 7 | 852 | ||
GenBank mRNA | AK044194.1 | - | 7 | 862 | ||
AK080814.1 | - | 7 | 850 | |||
AK144438.1 | - | 7 | 854 | |||
AK155061.1 | - | 7 | 867 | |||
AK184238.1 | - | 3 | 371 | |||
AK188222.1 | - | 3 | 357 | |||
BC018596.1 | - | 3 | 282 | |||
BC039649.1 | - | 7 | 846 | 5351885 | ||
D10715.1 | - | 7 | 852 |