Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007921.3 | NP_031947.1 | 2, 3 | 294 | ||
GenBank mRNA | AB221650.1 | - | 1, 2 | 162 | ||
AF016294.1 | - | 2, 3 | 260 | |||
BC138692.1 | - | 2, 3 | 258 | 8861714 |