Gene Symbol: ELOVL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022726.3 | NP_073563.1 | 2 | 531 | ||
GenBank mRNA | AF277094.1 | - | 2 | 359 | ||
AK055277.1 | - | 2 | 513 | |||
AK312511.1 | - | 2 | 486 | |||
AY037298.1 | - | 2 | 438 | |||
BC038506.1 | - | 2 | 376 | 5263012 |