Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159964.1 | NP_001153436.1 | 5, 6 | 987 | ||
NM_007943.3 | NP_031969.1 | 17, 18 | 1936 | |||
GenBank mRNA | AK028361.1 | - | 1135 | |||
AK083176.1 | - | 5, 6 | 971 | |||
AY190610.1 | - | 17, 18 | 1909 | |||
BC048783.1 | - | 17, 18 | 1910 | 6466448 | ||
L21768.1 | - | 17, 18 | 1909 |