Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007987.2 | NP_032013.2 | 9 | 769 | ||
GenBank mRNA | AK002590.1 | - | 9 | 720 | ||
BC061160.1 | - | 9 | 742 | 30302649 | ||
DQ846748.1 | - | 9 | 713 | |||
DQ846749.1 | - | 9 | 713 | |||
M83649.1 | - | 9 | 762 |