Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040403.1 | NP_001035493.1 | 3, 4 | 353 | ||
NM_001284227.1 | NP_001271156.1 | 4, 5 | 437 | |||
NM_001284228.1 | NP_001271157.1 | 4, 5 | 300 | |||
NM_008028.2 | NP_032054.1 | 4, 5 | 444 | |||
GenBank mRNA | AK004589.1 | - | 3, 4 | 291 | ||
AK030199.1 | - | 3, 4 | 316 | |||
AK149530.1 | - | 4, 5 | 375 | |||
AK170557.1 | - | 3, 4 | 354 | |||
AK191802.1 | - | 1, 2 | 57 | |||
AK213895.1 | - | 3, 4 | 208 | |||
BC070423.1 | - | 4, 5 | 402 | 6403816 | ||
BY204171.1 | - | 3, 4 | 354 | |||
U07890.1 | - | 4, 5 | 314 |