Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305115.1 | NP_001292044.1 | 2 | 591 | ||
NM_001305116.1 | NP_001292045.1 | 1 | 358 | |||
NM_024025.2 | NP_076930.1 | 2 | 594 | |||
GenBank mRNA | AB103376.1 | - | 2 | 409 | ||
AB158288.1 | - | 2 | 471 | |||
AB237597.1 | - | 2 | 406 | |||
AK055704.1 | - | 2 | 468 | |||
AY902194.1 | - | 2 | 409 | |||
BC001613.1 | - | 2 | 409 | 3535215 | ||
BC067804.1 | - | 2 | 261 | 5262839 | ||
BI753746.1 | - | 1 | 290 | |||
BI834891.1 | - | 1 | 359 | |||
BP196801.1 | - | 1 | 321 |