Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008052.3 | NP_032078.2 | 6 | 2082 | ||
GenBank mRNA | AB015422.1 | - | 5 | 1685 | ||
AK088630.1 | - | 6 | 2038 | |||
AK169455.1 | - | 6 | 2083 | |||
AK220514.1 | - | 5 | 1663 | |||
BC053055.1 | - | 6 | 1228 | 5685525 | ||
BC065075.1 | - | 7 | 1864 | |||
U38252.1 | - | 4 | 1267 |