Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113188.1 | NP_001106659.1 | 3, 4 | 240 | ||
NM_001113189.1 | NP_001106660.1 | 3, 4 | 240 | |||
NM_008053.2 | NP_032079.1 | 3, 4 | 240 | |||
GenBank mRNA | BC019139.1 | - | 3, 4 | 221 | 5041635 | |
CJ086949.1 | - | 3, 4 | 241 | |||
X90875.1 | - | 3, 4 | 219 |