Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6173
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
2
511
NM_001174093.1
NP_001167564.1
2
234
NM_001174094.1
NP_001167565.1
2
511
NM_001174096.1
NP_001167567.1
2
234
NM_001323638.1
NP_001310567.1
6
881
NM_001323641.1
NP_001310570.1
2
464
NM_001323642.1
NP_001310571.1
4
763
NM_001323643.1
NP_001310572.1
4
541
NM_001323645.1
NP_001310574.1
3
607
NM_001323646.1
NP_001310575.1
5
842
NM_001323647.1
NP_001310576.1
3
377
NM_001323648.1
NP_001310577.1
3
377
NM_001323649.1
NP_001310578.1
3
377
NM_001323650.1
NP_001310579.1
5
612
NM_001323651.1
NP_001310580.1
3
280
NM_001323652.1
NP_001310581.1
4
429
NM_001323653.1
NP_001310582.1
4
533
NM_001323654.1
NP_001310583.1
4
423
NM_001323655.1
NP_001310584.1
4
533
NM_001323656.1
NP_001310585.1
2
208
NM_001323657.1
NP_001310586.1
2
208
NM_001323659.1
NP_001310588.1
2
265
NM_001323660.1
NP_001310589.1
2
265
NM_001323661.1
NP_001310590.1
3
408
NM_001323662.1
NP_001310591.1
4
810
NM_001323663.1
NP_001310592.1
5
752
NM_001323664.1
NP_001310593.1
4
733
NM_001323665.1
NP_001310594.1
3
654
NM_001323666.1
NP_001310595.1
3
654
NM_001323671.1
NP_001310600.1
3
487
NM_001323672.1
NP_001310601.1
4
539
NM_001323673.1
NP_001310602.1
3
487
NM_001323674.1
NP_001310603.1
2
234
NM_001323675.1
NP_001310604.1
2
344
NM_001323676.1
NP_001310605.1
2
344
NM_001323677.1
NP_001310606.1
2
344
NM_001323678.1
NP_001310607.1
2
511
NM_030751.5
NP_110378.3
2
234
XM_006717498.2
XP_006717561.1
5
669
XM_011519643.2
XP_011517945.1
5
972
XM_017016597.1
XP_016872086.1
5
571
XM_017016598.1
XP_016872087.1
5
3701
XM_017016599.1
XP_016872088.1
4
3539
XM_017016600.1
XP_016872089.1
6
3780
XM_017016601.1
XP_016872090.1
5
3701
XM_017016602.1
XP_016872091.1
6
3780
XM_017016603.1
XP_016872092.1
5
3618
GenBank mRNA
AK091478.1
-
2
194
AK294775.1
-
2
429
AK296244.1
-
2
154
AK299666.1
-
2
265
AK307510.1
-
3
240
AK308785.1
-
3
319
AK308822.1
-
4
379
AK309331.1
-
4
763
AK310924.1
-
3
338
AK311116.1
-
2
247
AK311137.1
-
2
195
AK311187.1
-
2
494
AK311201.1
-
2
458
AK311372.1
-
5
788
AK311557.1
-
4
814
AK314683.1
-
2
195
AL831979.1
-
4
489
BC009572.1
-
2
190
BC065705.1
-
2
195
BC107781.1
-
2
190
BC112392.1
-
2
187
40036600
BX647794.1
-
6
882
BX954549.1
-
4
491
DB276952.1
-
4
389
DC299346.1
-
2
210
DC375708.1
-
2
265
U12170.1
-
2
195
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6d74b6fc5b-xzndl:80/10.239.164.225:80.
git-commit: 0683b6a9b3dadc77a268b704485bdfd9f1fd09b0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.0-2024.10.41-1.0