Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077411.2 | NP_001070879.1 | 8 | 1317 | ||
NM_008094.5 | NP_032120.1 | 9 | 1486 | |||
GenBank mRNA | AK082767.1 | - | 8 | 1315 | ||
AK152745.1 | - | 8 | 1298 | |||
AK166198.1 | - | 9 | 1477 | |||
AK205177.1 | - | 1 | 175 | |||
BC006663.1 | - | 9 | 1471 | 3498551 | ||
M24119.1 | - | 8 | 1226 |