Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008095.4 | NP_032121.3 | 5, 6 | 507 | ||
GenBank mRNA | AJ001261.1 | - | 5, 6 | 485 | ||
AK170336.1 | - | 5, 6 | 453 | |||
BC055893.1 | - | 5, 6 | 456 | 4023066 |