Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044744.1 | NP_001038209.2 | 11, 12 | 1461 | ||
NM_008097.2 | NP_032123.3 | 10, 11 | 1518 | |||
GenBank mRNA | AK078866.1 | - | 8, 9 | 1071 | ||
AK079500.1 | - | 4, 5 | 569 | |||
AK165406.1 | - | 10, 11 | 1344 | |||
BC061158.1 | - | 10, 11 | 1348 | 30314273 | ||
U18992.1 | - | 10, 11 | 1299 |