Gene Symbol: MIXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282402.1 | NP_001269331.1 | 2 | 697 | ||
NM_031944.2 | NP_114150.1 | 2 | 673 | |||
GenBank mRNA | AF211891.1 | - | 2 | 609 | ||
BC111974.1 | - | 2 | 673 | 8327442 | ||
BC113441.1 | - | 2 | 673 | 8322493 | ||
BC143784.1 | - | 2 | 697 | 9052298 |