Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008208.4 | NP_032234.3 | 6 | 2130 | ||
GenBank mRNA | AK033602.1 | - | 6 | 2131 |
Gene Symbol: H2-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153910.1 | - | 6 | 2058 |