Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Has2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008216.3 | NP_032242.3 | 2, 3 | 1188 | ||
GenBank mRNA | AK079729.1 | - | 2, 3 | 1181 | ||
BC080281.1 | - | 2, 3 | 1143 | 30533251 | ||
U52524.2 | - | 2, 3 | 1142 | |||
U69695.1 | - | 2, 3 | 1162 |