Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008152.3 | NP_032178.1 | 2 | 1549 | ||
GenBank mRNA | AK134113.1 | - | 2 | 1548 | ||
BC011332.1 | - | 1 | 1099 | 3661818 | ||
U39827.1 | - | 2 | 1527 |