Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008187.2 | NP_032213.2 | 3, 4 | 582 | ||
GenBank mRNA | AK011217.1 | - | 3, 4 | 526 | ||
AK088713.1 | - | 3, 4 | 486 | |||
AK172603.1 | - | 3, 4 | 558 | |||
BC092397.1 | - | 2, 3 | 2187 | |||
BC119023.1 | - | 3, 4 | 478 | 8733772 | ||
BC119025.1 | - | 3, 4 | 478 | 8733774 | ||
BC144770.1 | - | 2, 3 | 398 | |||
Z54179.1 | - | 3, 4 | 505 |