Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XYLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005108.3 | NP_005099.2 | 3 | 264 | ||
GenBank mRNA | AK025728.1 | - | 3 | 216 | ||
AK314386.1 | - | 3 | 212 | |||
BC030231.2 | - | 3 | 233 | |||
BC137076.1 | - | 3 | 196 | 9021073 | ||
BC137080.1 | - | 3 | 196 | 9021077 | ||
BI090475.1 | - | 3 | 127 |