Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxd13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008275.3 | NP_032301.2 | 1 | 578 | ||
GenBank mRNA | AB221620.1 | - | 1 | 578 | ||
AK077862.1 | - | 1 | 284 | |||
X99291.1 | - | 1 | 578 |