Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfals | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008340.3 | NP_032366.2 | 2 | 1027 | ||
GenBank mRNA | AK004926.1 | - | 2 | 1029 | ||
AK220290.1 | - | 2 | 1539 | |||
BC129875.1 | - | 2 | 1015 | 40110389 | ||
BC129876.1 | - | 2 | 1015 | 40110391 |