Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008269.1 | NP_032295.1 | 2 | 1139 | ||
GenBank mRNA | AK011622.1 | - | 2 | 1237 | ||
BC016893.1 | - | 2 | 1139 | 4223365 | ||
X56461.1 | - | 2 | 1652 |