Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008308.4 | NP_032334.2 | 1 | 4392 | ||
GenBank mRNA | AK049884.1 | - | 1 | 4324 | ||
BY601671.1 | - | 1 | 281 |
Set Membership: |
![]() |