Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Idh3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008323.1 | NP_032349.1 | 5, 6 | 395 | ||
GenBank mRNA | AJ748655.1 | - | 5, 6 | 347 | ||
AK156309.1 | - | 5, 6 | 393 | |||
AK166925.1 | - | 4, 5 | 352 | |||
AK167131.1 | - | 3, 4 | 923 | |||
AK168526.1 | - | 5, 6 | 393 | |||
BC085179.1 | - | 5, 6 | 368 | 30699000 | ||
BC119170.1 | - | 5, 6 | 378 | 8733919 | ||
BC119172.1 | - | 5, 6 | 378 | 8733921 | ||
U68564.1 | - | 5, 6 | 395 |