Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032408.3 | NP_115784.1 | 2 | 549 | ||
XM_017012773.1 | XP_016868262.1 | 2 | 562 | |||
GenBank mRNA | AB032253.1 | - | 2 | 542 | ||
AF072810.1 | - | 2 | 549 | |||
AF084479.1 | - | 2 | 481 | |||
BC065029.1 | - | 3 | 251 | |||
BC136520.1 | - | 2 | 382 | 9020510 |