Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008432.3 | NP_032458.3 | 1, 2 | 228 | ||
GenBank mRNA | AF039213.2 | - | 1, 2 | 195 | ||
CJ042506.1 | - | 1, 2 | 229 |