Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008410.2 | NP_032436.1 | 5, 6 | 868 | ||
GenBank mRNA | AB030203.1 | - | 5, 6 | 888 | ||
AB030204.1 | - | 4, 5 | 684 | |||
AK002599.1 | - | 5, 6 | 831 | |||
AK003224.1 | - | 5, 6 | 835 | |||
AK003587.1 | - | 5, 6 | 857 | |||
AK003928.1 | - | 5, 6 | 855 | |||
AK005125.1 | - | 5, 6 | 859 | |||
AK008861.1 | - | 1 | 23 | |||
AK076139.1 | - | 5, 6 | 877 | |||
AK077482.1 | - | 5, 6 | 870 | |||
AK150378.1 | - | 5, 6 | 888 | |||
AK151714.1 | - | 5, 6 | 887 | |||
AK151889.1 | - | 5, 6 | 888 | |||
AK152068.1 | - | 5, 6 | 876 | |||
AK152516.1 | - | 5, 6 | 888 | |||
AK152650.1 | - | 5, 6 | 888 | |||
AK152731.1 | - | 5, 6 | 859 | |||
AK152983.1 | - | 5, 6 | 880 | |||
AK153103.1 | - | 5, 6 | 831 | |||
AK159307.1 | - | 5, 6 | 832 | |||
AK159340.1 | - | 5, 6 | 856 | |||
AK159378.1 | - | 5, 6 | 887 | |||
AK159426.1 | - | 5, 6 | 856 | |||
AK159619.1 | - | 5, 6 | 880 | |||
AK159628.1 | - | 5, 6 | 856 | |||
AK159792.1 | - | 5, 6 | 856 | |||
AK159962.1 | - | 5, 6 | 859 | |||
AK160101.1 | - | 5, 6 | 856 | |||
AK160725.1 | - | 5, 6 | 887 | |||
AK200695.1 | - | 1, 2 | 91 | |||
BC004731.1 | - | 5, 6 | 839 | 3583154 | ||
BC010320.1 | - | 5, 6 | 871 | 3599190 | ||
BC021786.1 | - | 5, 6 | 877 | 4458918 | ||
U76253.1 | - | 5, 6 | 868 |