Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008428.4 | NP_032454.1 | 2, 3 | 629 | ||
GenBank mRNA | AK156346.1 | - | 2, 3 | 579 | ||
AK157219.1 | - | 1, 2 | 172 | |||
BC003756.1 | - | 2, 3 | 666 | 3490123 | ||
D88159.1 | - | 2, 3 | 585 |