Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008445.2 | NP_032471.2 | 1 | 2165 | ||
GenBank mRNA | AF013116.1 | - | 1 | 1741 | ||
AK133649.1 | - | 1 | 2147 | |||
AK147572.1 | - | 1 | 1967 | |||
AK181663.1 | - | 1 | 54 | |||
AK220283.1 | - | 2 | 1890 | |||
BC052732.1 | - | 1 | 1322 | |||
BC053053.1 | - | 1 | 2107 | |||
BC062648.1 | - | 1 | 1840 | |||
BC127063.1 | - | 1 | 1377 | 40109965 |