Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008451.2 | NP_032477.2 | 13 | 1776 | ||
GenBank mRNA | AF055666.1 | - | 13 | 1606 | ||
AK141439.1 | - | 13 | 1701 | |||
BC014845.1 | - | 13 | 1620 | 2599082 | ||
CX565742.1 | - | 5 | 376 |