Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZEB1 SIRNA Assays ›
Gene Symbol:
ZEB1
Gene Name:
zinc finger E-box binding homeobox 1
Gene Aliases:
AREB6, BZP, DELTAEF1, FECD6, NIL2A, PPCD3, TCF8, ZFHEP, ZFHX1A
Chromosome Location:
Chr.10: 31318495 - 31529814 on Build GRCh38
UniGene ID:
Hs.124503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6267
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZEB1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001128128.2
NP_001121600.1
2
560
NM_001174093.1
NP_001167564.1
2
283
NM_001174094.1
NP_001167565.1
2
560
NM_001174096.1
NP_001167567.1
2
283
NM_001323638.1
NP_001310567.1
6
930
NM_001323641.1
NP_001310570.1
2
513
NM_001323642.1
NP_001310571.1
4
812
NM_001323643.1
NP_001310572.1
4
590
NM_001323645.1
NP_001310574.1
3
656
NM_001323646.1
NP_001310575.1
5
891
NM_001323647.1
NP_001310576.1
3
426
NM_001323648.1
NP_001310577.1
3
426
NM_001323649.1
NP_001310578.1
3
426
NM_001323650.1
NP_001310579.1
5
661
NM_001323651.1
NP_001310580.1
3
329
NM_001323652.1
NP_001310581.1
4
478
NM_001323653.1
NP_001310582.1
4
582
NM_001323654.1
NP_001310583.1
4
472
NM_001323655.1
NP_001310584.1
4
582
NM_001323656.1
NP_001310585.1
2
257
NM_001323657.1
NP_001310586.1
2
257
NM_001323659.1
NP_001310588.1
2
314
NM_001323660.1
NP_001310589.1
2
314
NM_001323661.1
NP_001310590.1
3
457
NM_001323662.1
NP_001310591.1
4
859
NM_001323663.1
NP_001310592.1
5
801
NM_001323664.1
NP_001310593.1
4
782
NM_001323665.1
NP_001310594.1
3
703
NM_001323666.1
NP_001310595.1
3
703
NM_001323671.1
NP_001310600.1
3
536
NM_001323672.1
NP_001310601.1
4
588
NM_001323673.1
NP_001310602.1
3
536
NM_001323674.1
NP_001310603.1
2
283
NM_001323675.1
NP_001310604.1
2
393
NM_001323676.1
NP_001310605.1
2
393
NM_001323677.1
NP_001310606.1
2
393
NM_001323678.1
NP_001310607.1
2
560
NM_030751.5
NP_110378.3
2
283
XM_006717498.2
XP_006717561.1
5
718
XM_011519643.2
XP_011517945.1
5
1021
XM_017016597.1
XP_016872086.1
5
620
XM_017016598.1
XP_016872087.1
5
3750
XM_017016599.1
XP_016872088.1
4
3588
XM_017016600.1
XP_016872089.1
6
3829
XM_017016601.1
XP_016872090.1
5
3750
XM_017016602.1
XP_016872091.1
6
3829
XM_017016603.1
XP_016872092.1
5
3667
GenBank mRNA
AK091478.1
-
2
243
AK294775.1
-
2
478
AK296244.1
-
2
203
AK299666.1
-
2
314
AK307510.1
-
3
289
AK308785.1
-
3
368
AK308822.1
-
4
428
AK309331.1
-
4
812
AK309399.1
-
3
398
AK310924.1
-
3
387
AK311116.1
-
2
296
AK311137.1
-
2
244
AK311187.1
-
2
543
AK311201.1
-
2
507
AK311372.1
-
5
837
AK311557.1
-
4
863
AK314683.1
-
2
244
AL831979.1
-
4
538
BC009572.1
-
2
239
BC065705.1
-
2
244
BC107781.1
-
2
239
BC112392.1
-
2
236
40036600
BX647794.1
-
6
931
BX954549.1
-
4
541
DB276952.1
-
4
438
DC299346.1
-
2
259
DC375708.1
-
2
314
U12170.1
-
2
244
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
immune response
pattern specification process
central nervous system development
cell proliferation
negative regulation of cell proliferation
regulation of mesenchymal cell proliferation
regulation of transforming growth factor beta receptor signaling pathway
negative regulation of epithelial cell differentiation
regulation of T cell differentiation in thymus
positive regulation of neuron differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
embryonic camera-type eye morphogenesis
embryonic skeletal system morphogenesis
semicircular canal morphogenesis
regulation of smooth muscle cell differentiation
cartilage development
cellular response to amino acid stimulus
cochlea morphogenesis
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
transcription corepressor activity
protein binding
transcription factor binding
zinc ion binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0