Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008490.2 | NP_032516.2 | 1 | 149 | ||
GenBank mRNA | AI574112.1 | - | 1 | 149 | ||
AK004914.1 | - | 1 | 145 | |||
AK149476.1 | - | 1 | 146 | |||
BC028861.1 | - | 1 | 132 | 4212194 | ||
J05154.1 | - | 1 | 129 |