Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008508.2 | NP_032534.2 | 2 | 1628 | ||
GenBank mRNA | BC026781.1 | - | 2 | 1641 | ||
BC058223.1 | - | 2 | 1616 | 6307099 | ||
M34398.1 | - | 2 | 1611 |