Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198911.1 | NP_001185840.1 | 3 | 478 | ||
NM_001198912.1 | NP_001185841.1 | 2 | 340 | |||
NM_001198913.1 | NP_001185842.1 | 2 | 340 | |||
NM_008487.3 | NP_032513.3 | 2 | 350 | |||
GenBank mRNA | AB093254.1 | - | 2 | 423 | ||
AF177032.1 | - | 2 | 317 | |||
AK144972.1 | - | 3 | 459 | |||
AK156379.1 | - | 2 | 331 | |||
AK158232.1 | - | 3 | 543 | |||
AK167628.1 | - | 2 | 304 | |||
AK171223.1 | - | 2 | 299 | |||
BC006589.1 | - | 2 | 271 | 3587800 | ||
KC508090.1 | - | 2 | 326 | |||
U28495.1 | - | 2 | 317 | |||
X95761.1 | - | 2 | 208 |