Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145802.1 | NP_001139274.1 | 4, 5 | 619 | ||
NM_001145803.1 | NP_001139275.1 | 6 | 712 | |||
NM_008547.2 | NP_032573.2 | 5 | 607 | |||
GenBank mRNA | BC050009.1 | - | 6 | 651 | ||
BQ930470.1 | - | 6 | 627 | |||
X66983.1 | - | 5 | 601 |