Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008562.3 | NP_032588.1 | 2 | 875 | ||
GenBank mRNA | AK010424.1 | - | 1 | 50 | ||
AK159504.1 | - | 2 | 787 | |||
AK160594.1 | - | 1 | 47 | |||
AK215970.1 | - | 1 | 100 | |||
BC003839.1 | - | 2 | 804 | 3491015 | ||
BC005427.1 | - | 2 | 797 | 3493739 | ||
BC021638.1 | - | 2 | 814 | 4190996 | ||
GU182318.1 | - | 3 | 601 | |||
U35623.1 | - | 2 | 787 |