Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198911.1 | NP_001185840.1 | 13 | 1804 | ||
NM_001198912.1 | NP_001185841.1 | 12 | 1672 | |||
NM_001198913.1 | NP_001185842.1 | 12 | 1666 | |||
NM_008487.3 | NP_032513.3 | 12 | 1682 | |||
GenBank mRNA | AB093254.1 | - | 12 | 1755 | ||
AF177032.1 | - | 12 | 1649 | |||
AK144972.1 | - | 13 | 1785 | |||
AK156379.1 | - | 12 | 1664 | |||
AK158232.1 | - | 13 | 1875 | |||
AK167628.1 | - | 12 | 1636 | |||
AK171223.1 | - | 12 | 1625 | |||
BC006589.1 | - | 12 | 1603 | 3587800 | ||
U28495.1 | - | 12 | 1649 | |||
X95761.1 | - | 12 | 1540 |