Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008671.2 | NP_032697.2 | 1 | 468 | ||
GenBank mRNA | AK051439.1 | - | 1 | 218 | ||
AK077430.1 | - | 1 | 428 | |||
AK131621.1 | - | 1 | 346 | |||
BC115708.1 | - | 1 | 169 | 40058104 | ||
BC115761.1 | - | 1 | 169 | 40058103 | ||
X92352.1 | - | 1 | 444 |