Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008585.2 | NP_032611.2 | 12 | 1731 | ||
GenBank mRNA | AK093914.1 | - | 12 | 1690 | ||
AK136455.1 | - | 2 | 389 | |||
AK143906.1 | - | 12 | 1689 | |||
BC015258.1 | - | 12 | 1731 | 4241252 | ||
M74897.1 | - | 12 | 1730 |