Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008682.2 | NP_032708.2 | 3, 4 | 341 | ||
GenBank mRNA | AK010591.1 | - | 3, 4 | 329 | ||
AK048393.1 | - | 3, 4 | 293 | |||
AK085355.1 | - | 3, 4 | 304 | |||
AK090040.1 | - | 3, 4 | 286 | |||
AK153746.1 | - | 3, 4 | 343 | |||
BC052430.1 | - | 3, 4 | 310 | 6836583 | ||
BC066870.1 | - | 3, 4 | 278 | 30476383 | ||
D10712.1 | - | 3, 4 | 283 |