Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPVF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022150.3 | NP_071433.3 | 2, 3 | 594 | ||
GenBank mRNA | AB040290.1 | - | 2, 3 | 547 | ||
AF440392.1 | - | 2, 3 | 594 | |||
BM714303.1 | - | 2, 3 | 539 |