Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008723.1 | NP_032749.1 | 6 | 555 | ||
GenBank mRNA | AK011749.1 | - | 6 | 558 | ||
AK146603.1 | - | 6 | 558 | |||
AK157198.1 | - | 6 | 579 | |||
AK171848.1 | - | 6 | 558 | |||
BC048491.1 | - | 6 | 578 | 6704326 | ||
U64450.1 | - | 6 | 555 |