Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008855.2 | NP_032881.1 | 1 | 327 | ||
GenBank mRNA | AK051862.1 | - | 1 | 320 | ||
BC127083.1 | - | 1 | 146 | 40111744 | ||
BY729820.1 | - | 1 | 327 | |||
CJ050078.1 | - | 1 | 118 | |||
X53532.1 | - | 1 | 155 | |||
X59274.1 | - | 1 | 143 |