Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pemt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290011.1 | NP_001276940.1 | 7 | 754 | ||
NM_001290012.1 | NP_001276941.1 | 7 | 750 | |||
NM_001290013.1 | NP_001276942.1 | 7 | 688 | |||
NM_001290014.1 | NP_001276943.1 | 5 | 430 | |||
NM_008819.3 | NP_032845.2 | 7 | 788 | |||
GenBank mRNA | AA245746.1 | - | 32 | |||
AK133744.1 | - | 7 | 750 | |||
BC026796.1 | - | 7 | 622 | 4196324 |